Erythrosin B, pure, high purity, biological stain
Erythrosin B, pure, high purity, biological stain
Erythrosin B, pure, high purity, biological stain
Erythrosin B, pure, high purity, biological stain
Thermo Scientific Chemicals
Erythrosin B, pure, high purity, biological stain
CAS: 16423-68-0 | C20H6I4Na2O5 | 879.86 g/mol
Have Questions?
Catalog NumberQuantity
ACR18946025025 g
Catalog number ACR189460250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS7440-43-9
IUPAC Namecadmium(2+) ion
Molecular FormulaCd
InChI KeyWLZRMCYVCSSEQC-UHFFFAOYSA-N
SMILES[Cd++]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White silvery
FormFoil, 1.0mm (0.04in) thick.
Assay from Supplier's CofA≥99.95% (metals basis)
Width15cm (5.9in) width.
Indicator
RUO – Research Use Only