DL-10-Camphorsulfonic acid, 98%, Thermo Scientific Chemicals
DL-10-Camphorsulfonic acid, 98%, Thermo Scientific Chemicals
DL-10-Camphorsulfonic acid, 98%, Thermo Scientific Chemicals
DL-10-Camphorsulfonic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
DL-10-Camphorsulfonic acid, 98%, Thermo Scientific Chemicals
CAS: 5872-08-2 | C10H16O4S | 232.29 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Plastic bottle
Catalog number ACR189071000
Price (MYR)
575.00
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
575.00
Add to cart
Specifications
Chemical Name or MaterialDL-10-Camphorsulfonic acid
Name Note98%
CAS5872-08-2
Absorbance(20% aq. soln.)
Health Hazard 1GHS Signal Word: Danger