3-Fluoropyridine, 99%, Thermo Scientific Chemicals
3-Fluoropyridine, 99%, Thermo Scientific Chemicals
3-Fluoropyridine, 99%, Thermo Scientific Chemicals
3-Fluoropyridine, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
3-Fluoropyridine, 99%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number ACR188300050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS501-52-0
IUPAC Name3-phenylpropanoic acid
Molecular FormulaC9H10O2
InChI KeyXMIIGOLPHOKFCH-UHFFFAOYSA-N
SMILESOC(=O)CCC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals or mass and/or
Infrared spectrumConforms
GC>=98.5 %
Appearance (Color)White to yellow to light beige
Appearance (Form)chunks