Levamisole hydrochloride, 99+%
Levamisole hydrochloride, 99+%
Levamisole hydrochloride, 99+%
Levamisole hydrochloride, 99+%
Thermo Scientific Chemicals
Levamisole hydrochloride, 99+%
CAS: 16595-80-5 | C11H12N2S·ClH | 240.76 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
Packaging:
Catalog number ACR187870100
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Request bulk or custom format
Chemical Identifiers
CAS7440-22-4
IUPAC Namesilver
Molecular FormulaAg
InChI KeyBQCADISMDOOEFD-UHFFFAOYSA-N
SMILES[Ag]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Silver
FormWire, 0.05mm (0.002in) diameter, annealed.
Assay from Supplier's CofA≥99.99% (metals basis)