2,2-Dimethoxy-2-phenylacetophenone, 99%, Thermo Scientific Chemicals
2,2-Dimethoxy-2-phenylacetophenone, 99%, Thermo Scientific Chemicals
2,2-Dimethoxy-2-phenylacetophenone, 99%, Thermo Scientific Chemicals
2,2-Dimethoxy-2-phenylacetophenone, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2,2-Dimethoxy-2-phenylacetophenone, 99%, Thermo Scientific Chemicals
CAS: 24650-42-8 | C16H16O3 | 256.30 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
5 g
50 g
Packaging:
Glass bottle
Catalog number ACR187842500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS14320-04-8
IUPAC Namezinc(2+) 2,11,20,29,37,38,39,40-octaazanonacyclo[28.6.1.1³,¹⁰.1¹²,¹⁹.1²¹,²⁸.0⁴,⁹.0¹³,¹⁸.0²²,²⁷.0³¹,³⁶]tetraconta-1(36),2,4,6,8,10(40),11,13,15,17,19(39),20,22,24,26,28,30,32,34-nonadecaene-37,38-diide
Molecular FormulaC32H16N8Zn
InChI KeyPODBBOVVOGJETB-UHFFFAOYSA-N
SMILES[Zn++].[N-]1C2=NC3=C4C=CC=CC4=C([N-]3)N=C3N=C(N=C4N=C(N=C1C1=CC=CC=C21)C1=CC=CC=C41)C1=CC=CC=C31
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
UV spectrophotometry>=95 %
Loss on drying=<5 % (105°C)
Appearance (Color)Blue to purple
Appearance (Form)Powder