2,2-Dimethoxy-2-phenylacetophenone, 99%, Thermo Scientific Chemicals
2,2-Dimethoxy-2-phenylacetophenone, 99%, Thermo Scientific Chemicals
2,2-Dimethoxy-2-phenylacetophenone, 99%, Thermo Scientific Chemicals
2,2-Dimethoxy-2-phenylacetophenone, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2,2-Dimethoxy-2-phenylacetophenone, 99%, Thermo Scientific Chemicals
CAS: 24650-42-8 | C16H16O3 | 256.30 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
50 g
250 g
Packaging:
Glass bottle
Catalog number ACR187840050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material2, 2-Dimethoxy-2-phenylacetophenone
Name Note99%
CAS24650-42-8
CAS Min %98.5
Health Hazard 1LIGHT SENSITIVE