4-O-Methyldopamine hydrochloride, 98%, Thermo Scientific Chemicals
4-O-Methyldopamine hydrochloride, 98%, Thermo Scientific Chemicals
4-O-Methyldopamine hydrochloride, 98%, Thermo Scientific Chemicals
4-O-Methyldopamine hydrochloride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-O-Methyldopamine hydrochloride, 98%, Thermo Scientific Chemicals
CAS: 645-33-0 | C9H13NO2·HCl | 203.67 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mg
500 mg
Packaging:
Glass bottle
Catalog number ACR187740500
Price (MYR)
560.00
Add to cart
Quantity:
50 mg
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
560.00
Add to cart
Specifications
Chemical Name or Material4-O-Methyldopamine hydrochloride
CAS645-33-0
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0