2-Nitroimidazole, 98%, Thermo Scientific Chemicals
2-Nitroimidazole, 98%, Thermo Scientific Chemicals
2-Nitroimidazole, 98%, Thermo Scientific Chemicals
2-Nitroimidazole, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Nitroimidazole, 98%, Thermo Scientific Chemicals
CAS: 527-73-1 | C3H3N3O2 | 113.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
Packaging:
Glass bottle
Catalog number ACR187572500
Price (MYR)
364.83
Add to cart
Quantity:
250 mg
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
364.83
Add to cart
Specifications
Chemical Name or Material2-Nitroimidazole
Name Note98%
CAS527-73-1
CAS Min %97.5
Health Hazard 1GHS Signal Word: Danger