1-Bromotetradecane, 98%, Thermo Scientific Chemicals
1-Bromotetradecane, 98%, Thermo Scientific Chemicals
1-Bromotetradecane, 98%, Thermo Scientific Chemicals
1-Bromotetradecane, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Bromotetradecane, 98%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Glass bottle
Catalog number ACR187381000
Price (MYR)
609.00
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
609.00
Add to cart
Specifications
Chemical Name or Material1-Bromotetradecane
Name Note98%
CAS112-71-0
CAS Min %97.5
CAS Max %100.0