O-(2,3,4,5,6-Pentafluorobenzyl)hydroxylamine hydrochloride, 98%, Thermo Scientific Chemicals
O-(2,3,4,5,6-Pentafluorobenzyl)hydroxylamine hydrochloride, 98%, Thermo Scientific Chemicals
O-(2,3,4,5,6-Pentafluorobenzyl)hydroxylamine hydrochloride, 98%, Thermo Scientific Chemicals
O-(2,3,4,5,6-Pentafluorobenzyl)hydroxylamine hydrochloride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
O-(2,3,4,5,6-Pentafluorobenzyl)hydroxylamine hydrochloride, 98%, Thermo Scientific Chemicals
O-(2,3,4,5,6-Pentafluorobenzyl)hydroxylamine hydrochloride, 98%, C7H4F5NO.HCl, CAS Number-57981-02-9
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Catalog number ACR186710050
Price (MYR)
4,599.00
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
4,599.00
Add to cart
Chemical Identifiers
CAS1310-58-3
IUPAC Namepotassium hydroxide
Molecular FormulaHKO
InChI KeyKWYUFKZDYYNOTN-UHFFFAOYSA-M
SMILES[OH-].[K+]
View more
SpecificationsSpecification SheetSpecification Sheet
Strength0.950 to 1.050 N
Appearance (Color)Clear colorless to yellow to brown
Appearance (Form)Solution