Dimethyl isophthalate, 99%, Thermo Scientific Chemicals
Dimethyl isophthalate, 99%, Thermo Scientific Chemicals
Dimethyl isophthalate, 99%, Thermo Scientific Chemicals
Dimethyl isophthalate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Dimethyl isophthalate, 99%, Thermo Scientific Chemicals
CAS: 1459-93-4 | C10H10O4 | 194.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
Packaging:
Plastic bottle
Catalog number ACR186510025
Price (MYR)
1,308.00
BOT
Add to cart
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
1,308.00
BOT
Add to cart
Chemical Identifiers
CAS9010-66-6
MDL NumberMFCD00062414
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to straw yellow to tan
Infrared spectrumConforms
Loss on drying=<8.0 % (105°C, 2 h)
Appearance (Form)Powder
Residue after ignition=<2.0 % (800°C)