Oxalic acid, 98%, anhydrous, Thermo Scientific Chemicals
Oxalic acid, 98%, anhydrous, Thermo Scientific Chemicals
Oxalic acid, 98%, anhydrous, Thermo Scientific Chemicals
Oxalic acid, 98%, anhydrous, Thermo Scientific Chemicals
Oxalic acid, 98%, anhydrous, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Oxalic acid, 98%, anhydrous, Thermo Scientific Chemicals
CAS: 144-62-7 | C2H2O4 | 90.034 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Packaging:
View all
Glass bottle
Catalog number ACR186432500
Price (MYR)
479.00
Add to cart
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
479.00
Add to cart
Chemical Identifiers
CAS13889-98-0
IUPAC Name4-acetylpiperazin-1-ium
Molecular FormulaC6H13N2O
InChI KeyPKDPUENCROCRCH-UHFFFAOYSA-O
SMILESCC(=O)N1CC[NH2+]CC1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥98.5%
Appearance (Color)White to cream to yellow to green
Melting Point (clear melt)28.0-35.0?C
FormCrystals or powder or crystalline powder or fused solid or liquid as melt
Suitable for battery materials development.
RUO – Research Use Only