1,3-Dimethyl-2-imidazolidinone, 98%, Thermo Scientific Chemicals
1,3-Dimethyl-2-imidazolidinone, 98%, Thermo Scientific Chemicals
1,3-Dimethyl-2-imidazolidinone, 98%, Thermo Scientific Chemicals
1,3-Dimethyl-2-imidazolidinone, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1,3-Dimethyl-2-imidazolidinone, 98%, Thermo Scientific Chemicals
CAS: 80-73-9 | C5H10N2O | 114.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
100 g
Packaging:
Glass bottle
Catalog number ACR185930010
Price (MYR)
1,959.00
Add to cart
Quantity:
1 kg
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,959.00
Add to cart
Specifications
Chemical Name or Material1,3-Dimethyl-2-imidazolidinone
Name Note98%
CAS80-73-9
CAS Min %97.5
Health Hazard 1GHS Signal Word: Danger