Ethylaluminium dichloride, 0.9M solution in heptane, AcroSeal™, Thermo Scientific Chemicals
Ethylaluminium dichloride, 0.9M solution in heptane, AcroSeal™, Thermo Scientific Chemicals
Ethylaluminium dichloride, 0.9M solution in heptane, AcroSeal™, Thermo Scientific Chemicals
Ethylaluminium dichloride, 0.9M solution in heptane, AcroSeal™, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Ethylaluminium dichloride, 0.9M solution in heptane, AcroSeal™, Thermo Scientific Chemicals
CAS: 563-43-9 | C2H5AlCl2 | 126.94 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
Packaging:
AcroSeal™ Glass bottle
Catalog number ACR185451000
Price (MYR)
950.00
Add to cart
Quantity:
100 mL
Packaging:
AcroSeal™ Glass bottle
Request bulk or custom format
Price (MYR)
950.00
Add to cart
Chemical Identifiers
CAS75-66-1
IUPAC Name2-methylpropane-2-thiol
Molecular FormulaC4H10S
InChI KeyWMXCDAVJEZZYLT-UHFFFAOYSA-N
SMILESCC(C)(C)S
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=98.5 %
Refractive index1.4220 to 1.4240 (20°C, 589 nm)
Lewis acid
RUO – Research Use Only