Poly(acrylamide), carboxyl modified, pure, high carboxyl content, ∽ MW 200,000, Thermo Scientific™
Poly(acrylamide), carboxyl modified, pure, high carboxyl content, ∽ MW 200,000, Thermo Scientific™
Poly(acrylamide), carboxyl modified, pure, high carboxyl content, ∽ MW 200,000, Thermo Scientific™
Poly(acrylamide), carboxyl modified, pure, high carboxyl content, ∽ MW 200,000, Thermo Scientific™
Poly(acrylamide), carboxyl modified, pure, high carboxyl content, ∽ MW 200,000, Thermo Scientific™
Have Questions?
Catalog number ACR184240050
Price (MYR)
1.00
Add to cart
Request bulk or custom format
Price (MYR)
1.00
Add to cart
Specifications
Chemical Name or MaterialPoly(acrylamide), carboxyl modified
ColorWhite
CAS62649-23-4
Physical FormGranules
Molecular Formula(C3H5NO)n