1H-Benzotriazole, 99%, Thermo Scientific Chemicals
1H-Benzotriazole, 99%, Thermo Scientific Chemicals
1H-Benzotriazole, 99%, Thermo Scientific Chemicals
1H-Benzotriazole, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1H-Benzotriazole, 99%, Thermo Scientific Chemicals
CAS: 95-14-7 | C6H5N3 | 119.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Catalog number ACR183881000
Price (MYR)
524.00
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
524.00
Add to cart
Chemical Identifiers
CAS141-12-8
IUPAC Name(2Z)-3,7-dimethylocta-2,6-dien-1-yl acetate
Molecular FormulaC12H20O2
InChI KeyHIGQPQRQIQDZMP-FLIBITNWSA-N
SMILESCC(C)=CCCC(C)=C/COC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
Appearance (Color)Clear colorless
Assay (GC)≥97.5%
Refractive Index1.4590-1.4620 @ 20?C
FormLiquid
Indicator
RUO – Research Use Only