1H-Benzotriazole, 99%, Thermo Scientific Chemicals
1H-Benzotriazole, 99%, Thermo Scientific Chemicals
1H-Benzotriazole, 99%, Thermo Scientific Chemicals
1H-Benzotriazole, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1H-Benzotriazole, 99%, Thermo Scientific Chemicals
CAS: 95-14-7 | C6H5N3 | 119.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
100 g
500 g
Packaging:
Plastic bottle
Catalog number ACR183880025
Price (MYR)
3,071.00
Add to cart
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
3,071.00
Add to cart
Chemical Identifiers
CAS21739-92-4
IUPAC Name5-bromo-2-chlorobenzoate
Molecular FormulaC7H3BrClO2
InChI KeyFGERXQWKKIVFQG-UHFFFAOYSA-M
SMILES[O-]C(=O)C1=CC(Br)=CC=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)≥98.0 to ≤102.0%
Assay (Silylated GC)≥98.0%
Melting Point (clear melt)155.0-162.0?C
Appearance (Color)White
Indicator
RUO – Research Use Only