3-Phenoxybenzyl alcohol, 98%, Thermo Scientific Chemicals
3-Phenoxybenzyl alcohol, 98%, Thermo Scientific Chemicals
3-Phenoxybenzyl alcohol, 98%, Thermo Scientific Chemicals
3-Phenoxybenzyl alcohol, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
3-Phenoxybenzyl alcohol, 98%, Thermo Scientific Chemicals
CAS: 13826-35-2 | C13H12O2 | 200.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number ACR183770250
Price (MYR)
846.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
846.00
Add to cart
Chemical Identifiers
CAS7791-18-6
IUPAC Namemagnesium(2+) hexahydrate dichloride
Molecular FormulaCl2H12MgO6
InChI KeyDHRRIBDTHFBPNG-UHFFFAOYSA-L
SMILESO.O.O.O.O.O.[Mg++].[Cl-].[Cl-]
View more
SpecificationsSpecification SheetSpecification Sheet
CommentSulfate: ≤0.002%
Heavy Metals≤5 ppm (as Pb)
Assay from Supplier's CofA≥99.0% (titration)
CommentAmmonium: ≤0.002%
CommentBarium: ≤0.005%