Poly(vinyl alcohol), 98.0%-98.8% hydrolyzed, M.W. approx. 50,000-85,000, Thermo Scientific Chemicals
Poly(vinyl alcohol), 98.0%-98.8% hydrolyzed, M.W. approx. 50,000-85,000, Thermo Scientific Chemicals
Poly(vinyl alcohol), 98.0%-98.8% hydrolyzed, M.W. approx. 50,000-85,000, Thermo Scientific Chemicals
Poly(vinyl alcohol), 98.0%-98.8% hydrolyzed, M.W. approx. 50,000-85,000, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Poly(vinyl alcohol), 98.0%-98.8% hydrolyzed, M.W. approx. 50,000-85,000, Thermo Scientific Chemicals
Indicator
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
1 kg
Catalog number ACR183381000
Price (MYR)
411.00
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
411.00
Add to cart
Specifications
Chemical Name or MaterialPoly(vinyl alcohol)
CAS9002-89-5
Melting Point230.0°C to 240.0°C
pH5.0 to 7.0 (4% aq. soln.)
Infrared SpectrumAuthentic