15-Crown-5, 98%, Thermo Scientific Chemicals
15-Crown-5, 98%, Thermo Scientific Chemicals
15-Crown-5, 98%, Thermo Scientific Chemicals
15-Crown-5, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
15-Crown-5, 98%, Thermo Scientific Chemicals
CAS: 33100-27-5 | C10H20O5 | 220.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number ACR182860250
Price (MYR)
905.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
905.00
Add to cart
Specifications
Chemical Name or Material15-Crown-5
Name Note98%
CAS33100-27-5
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning
View more
Chelation/Complexation reagent
RUO – Research Use Only