Cyclobutanemethanol, 99%, Thermo Scientific Chemicals
Cyclobutanemethanol, 99%, Thermo Scientific Chemicals
Cyclobutanemethanol, 99%, Thermo Scientific Chemicals
Cyclobutanemethanol, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Cyclobutanemethanol, 99%, Thermo Scientific Chemicals
CAS: 4415-82-1 | C5H10O | 86.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number ACR182230250
Price (MYR)
1,383.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,383.00
Add to cart
Chemical Identifiers
CAS693-58-3
IUPAC Name1-bromononane
Molecular FormulaC9H19Br
InChI KeyAYMUQTNXKPEMLM-UHFFFAOYSA-N
SMILESCCCCCCCCCBr
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
Refractive Index1.4530-1.4560 @ 20?C
Assay (GC)≥98.5%