3-Bromobenzyl bromide, 95%, Thermo Scientific Chemicals
3-Bromobenzyl bromide, 95%, Thermo Scientific Chemicals
3-Bromobenzyl bromide, 95%, Thermo Scientific Chemicals
3-Bromobenzyl bromide, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
3-Bromobenzyl bromide, 95%, Thermo Scientific Chemicals
CAS: 823-78-9 | C7H6Br2 | 249.93 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number ACR181780050
Price (MYR)
408.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
408.00
Add to cart
Specifications
Chemical Name or Material3-Bromobenzyl bromide
Name Note95%
CAS823-78-9
CAS Min %94.0
Health Hazard 1GHS Signal Word: Danger