5-Aminoisophthalic acid, 98%, Thermo Scientific Chemicals
5-Aminoisophthalic acid, 98%, Thermo Scientific Chemicals
5-Aminoisophthalic acid, 98%, Thermo Scientific Chemicals
5-Aminoisophthalic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
5-Aminoisophthalic acid, 98%, Thermo Scientific Chemicals
CAS: 99-31-0 | C8H7NO4 | 181.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Plastic bottle
Catalog number ACR181331000
Price (MYR)
954.00
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
954.00
Add to cart
Chemical Identifiers
CAS10025-65-7
IUPAC Nameplatinum(2+) dichloride
Molecular FormulaCl2Pt
InChI KeyCLSUSRZJUQMOHH-UHFFFAOYSA-L
SMILES[Cl-].[Cl-].[Pt++]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Appearance (Color)Grey-green to brown
Assay>=71.8 % (Pt)