5-Aminoisophthalic acid, 98%, Thermo Scientific Chemicals
5-Aminoisophthalic acid, 98%, Thermo Scientific Chemicals
5-Aminoisophthalic acid, 98%, Thermo Scientific Chemicals
5-Aminoisophthalic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
5-Aminoisophthalic acid, 98%, Thermo Scientific Chemicals
CAS: 99-31-0 | C8H7NO4 | 181.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Plastic bottle
Catalog number ACR181330250
Price (MYR)
461.00
Add to cart
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
461.00
Add to cart
Specifications
Chemical Name or Material5-Aminoisophthalic acid
Name Note98+%
CAS99-31-0
CAS Min %98.0
Health Hazard 1GHS Signal Word: Warning