N,N-Dimethylacetamide, 99+%, extra pure, Thermo Scientific Chemicals
N,N-Dimethylacetamide, 99+%, extra pure, Thermo Scientific Chemicals
N,N-Dimethylacetamide, 99+%, extra pure, Thermo Scientific Chemicals
N,N-Dimethylacetamide, 99+%, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
N,N-Dimethylacetamide, 99+%, extra pure, Thermo Scientific Chemicals
CAS: 127-19-5 | C4H9NO | 87.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 L
1 L
2.5 L
Packaging:
View all
Plastic Drum
Catalog number ACR181070050
Price (MYR)
1,215.00
Add to cart
Quantity:
5 L
Packaging:
Plastic Drum
Request bulk or custom format
Price (MYR)
1,215.00
Add to cart
Chemical Identifiers
CAS103-29-7
IUPAC Name(2-phenylethyl)benzene
Molecular FormulaC14H14
InChI KeyQWUWMCYKGHVNAV-UHFFFAOYSA-N
SMILESC(CC1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Infrared spectrumConforms
GC>=98.5 %
Appearance (Form)Powder or crystals and/or chunks
Melting point49°C to 53°C