N,N-Dimethylacetamide, 99+%, extra pure, Thermo Scientific Chemicals
N,N-Dimethylacetamide, 99+%, extra pure, Thermo Scientific Chemicals
N,N-Dimethylacetamide, 99+%, extra pure, Thermo Scientific Chemicals
N,N-Dimethylacetamide, 99+%, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
N,N-Dimethylacetamide, 99+%, extra pure, Thermo Scientific Chemicals
CAS: 127-19-5 | C4H9NO | 87.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
5 L
Packaging:
View all
Glass Bottle
Catalog number ACR181070025
Price (MYR)
980.00
Add to cart
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
980.00
Add to cart
Specifications
Chemical Name or MaterialN, N-Dimethylacetamide
Melting Point-20.0°C
CAS127-19-5
CAS Min %99.0
Health Hazard 1Signal Word: WARNING!