N,N-Dimethylacetamide, 99+%, extra pure, Thermo Scientific Chemicals
N,N-Dimethylacetamide, 99+%, extra pure, Thermo Scientific Chemicals
N,N-Dimethylacetamide, 99+%, extra pure, Thermo Scientific Chemicals
N,N-Dimethylacetamide, 99+%, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
N,N-Dimethylacetamide, 99+%, extra pure, Thermo Scientific Chemicals
CAS: 127-19-5 | C4H9NO | 87.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
5 L
Packaging:
View all
Glass Bottle
Catalog number ACR181070010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS79-42-5
IUPAC Name2-sulfanylpropanoic acid amine
Molecular FormulaC3H9NO2S
InChI KeyVHFSMDHANDABQK-UHFFFAOYNA-N
SMILESN.CC(S)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥96.0%
FormLiquid
Refractive Index1.4795-1.4845 @ 20?C
Appearance (Color)Clear colorless
Assay (Aqueous acid-base Titration)≥96.0 to ≤104.0%