Dicyclohexylamine, 99+%, Thermo Scientific Chemicals
Dicyclohexylamine, 99+%, Thermo Scientific Chemicals
Dicyclohexylamine, 99+%, Thermo Scientific Chemicals
Dicyclohexylamine, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Dicyclohexylamine, 99+%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
2.5 L
Packaging:
Glass bottle
Catalog number ACR181051000
Price (MYR)
416.00
Add to cart
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
416.00
Add to cart
Chemical Identifiers
CAS40615-36-9
IUPAC Name1-[chloro(4-methoxyphenyl)phenylmethyl]-4-methoxybenzene
Molecular FormulaC21H19ClO2
InChI KeyJBWYRBLDOOOJEU-UHFFFAOYSA-N
SMILESCOC1=CC=C(C=C1)C(Cl)(C1=CC=CC=C1)C1=CC=C(OC)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale pink to orange
Assay (Titration ex Chloride)≥96.0 to ≤104.0%
FormPowder
Assay (HPLC)≥96.0%