Dicyclohexylamine, 99+%, Thermo Scientific Chemicals
Dicyclohexylamine, 99+%, Thermo Scientific Chemicals
Dicyclohexylamine, 99+%, Thermo Scientific Chemicals
Dicyclohexylamine, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Dicyclohexylamine, 99+%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
100 mL
Packaging:
Glass bottle
Catalog number ACR181050025
Price (MYR)
717.00
Add to cart
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
717.00
Add to cart
Chemical Identifiers
CAS10108-64-2
IUPAC Namecadmium(2+) dichloride
Molecular FormulaCdCl2
InChI KeyYKYOUMDCQGMQQO-UHFFFAOYSA-L
SMILES[Cl-].[Cl-].[Cd++]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.004% max.
Appearance (Color)White
Residual water< 100 ppm