Cyanuric acid, 98%, Thermo Scientific Chemicals
Cyanuric acid, 98%, Thermo Scientific Chemicals
Cyanuric acid, 98%, Thermo Scientific Chemicals
Cyanuric acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Cyanuric acid, 98%, Thermo Scientific Chemicals
CAS: 108-80-5 | C3H3N3O3 | 129.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
250 g
1 kg
Packaging:
Plastic bottle
Catalog number ACR181020250
Price (MYR)
350.00
Add to cart
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
350.00
Add to cart
Chemical Identifiers
CAS2167-23-9
IUPAC Name2,2-bis(tert-butylperoxy)butane
Molecular FormulaC12H26O4
InChI KeyHQOVXPHOJANJBR-UHFFFAOYSA-N
SMILESCCC(C)(OOC(C)(C)C)OOC(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Color scale=<10 APHA
Titration Iodometric49 to 51 %
Impurity=<0.3 % Hydroperoxides (as TBHP)
Appearance (Form)Clear solution