Cyanuric acid, 98%, Thermo Scientific Chemicals
Cyanuric acid, 98%, Thermo Scientific Chemicals
Cyanuric acid, 98%, Thermo Scientific Chemicals
Cyanuric acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Cyanuric acid, 98%, Thermo Scientific Chemicals
CAS: 108-80-5 | C3H3N3O3 | 129.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
25 g
250 g
Packaging:
Plastic bottle
Catalog number ACR181020010
Price (MYR)
1,586.00
Add to cart
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
1,586.00
Add to cart
Chemical Identifiers
CAS696-60-6
IUPAC Name4-(aminomethyl)phenol
Molecular FormulaC7H9NO
InChI KeyRQJDUEKERVZLLU-UHFFFAOYSA-N
SMILESNCC1=CC=C(O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Elemental Analysis57.18 to 61.94 % (C) based on monohydrate
Appearance (Color)Off-white to light yellow
Infrared spectrumConforms
Elemental Analysis9.52 to 10.31 % (N) based on monohydrate