Cumene, 99%, pure, Thermo Scientific Chemicals
Cumene, 99%, pure, Thermo Scientific Chemicals
Cumene, 99%, pure, Thermo Scientific Chemicals
Cumene, 99%, pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Cumene, 99%, pure, Thermo Scientific Chemicals
CAS: 98-82-8 | C9H12 | 120.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Packaging:
Glass bottle
Catalog number ACR181010025
Price (MYR)
1,040.00
Add to cart
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,040.00
Add to cart
Chemical Identifiers
CAS2408-37-9
IUPAC Name2,2,6-trimethylcyclohexan-1-one
Molecular FormulaC9H16O
InChI KeyZPVOLGVTNLDBFI-UHFFFAOYNA-N
SMILESCC1CCCC(C)(C)C1=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Appearance (Color)Clear colorless to pale yellow
Identification (FTIR)Conforms
Refractive Index1.4450-1.4500 @ 20?C
Assay (GC)≥96.0%