Cumene, 99%, pure, Thermo Scientific Chemicals
Cumene, 99%, pure, Thermo Scientific Chemicals
Cumene, 99%, pure, Thermo Scientific Chemicals
Cumene, 99%, pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Cumene, 99%, pure, Thermo Scientific Chemicals
CAS: 98-82-8 | C9H12 | 120.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Packaging:
Glass bottle
Catalog number ACR181010010
Price (MYR)
583.00
Add to cart
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
583.00
Add to cart
Chemical Identifiers
CAS96-47-9
IUPAC Name2-methyloxolane
Molecular FormulaC5H10O
InChI KeyJWUJQDFVADABEY-UHFFFAOYNA-N
SMILESCC1CCCO1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Peroxides=<50 ppm
Infrared spectrumConforms
GC>=99.0 %
Appearance (Form)Liquid