Naphthalene, 99%, Thermo Scientific Chemicals
Naphthalene, 99%, Thermo Scientific Chemicals
Naphthalene, 99%, Thermo Scientific Chemicals
Naphthalene, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Naphthalene, 99%, Thermo Scientific Chemicals
CAS: 91-20-3 | C10H8 | 128.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 kg
250 g
1 kg
2.5 kg
Packaging:
View all
Plastic drum
Catalog number ACR180900050
Price (MYR)
1,222.00
Add to cart
Quantity:
5 kg
Packaging:
Plastic drum
Request bulk or custom format
Price (MYR)
1,222.00
Add to cart
Chemical Identifiers
CAS67-64-1
IUPAC Namepropan-2-one
Molecular FormulaC3H6O
InChI KeyCSCPPACGZOOCGX-UHFFFAOYSA-N
SMILESCC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Color scale=<10 APHA
Water=<0.005 % (Coulometric)
Appearance (Form)Clear liquid
Acidity (CH3COOH)=<0.003 %
GC>=99.75 %