Naphthalene, 99%, Thermo Scientific Chemicals
Naphthalene, 99%, Thermo Scientific Chemicals
Naphthalene, 99%, Thermo Scientific Chemicals
Naphthalene, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Naphthalene, 99%, Thermo Scientific Chemicals
CAS: 91-20-3 | C10H8 | 128.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
250 g
1 kg
5 kg
Packaging:
View all
Plastic bottle
Catalog number ACR180900025
Price (MYR)
811.00
Add to cart
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
811.00
Add to cart
Chemical Identifiers
CAS10024-93-8
IUPAC Nameneodymium(3+) trichloride
Molecular FormulaCl3Nd
InChI KeyATINCSYRHURBSP-UHFFFAOYSA-K
SMILES[Cl-].[Cl-].[Cl-].[Nd+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Grey to blue to purple
FormCrystals or powder or crystalline powder or fused or granular solid
Assay (unspecified)Total rare earth chlorides: ≥99.0%
Residual water≤1.0%
Impurity contentCalcium oxide, CaO: ≤0.01% (≤100ppm)