Naphthalene, 99%, Thermo Scientific Chemicals
Naphthalene, 99%, Thermo Scientific Chemicals
Naphthalene, 99%, Thermo Scientific Chemicals
Naphthalene, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Naphthalene, 99%, Thermo Scientific Chemicals
CAS: 91-20-3 | C10H8 | 128.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
250 g
2.5 kg
5 kg
Packaging:
View all
Plastic bottle
Catalog number ACR180900010
Price (MYR)
556.00
Add to cart
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
556.00
Add to cart
Chemical Identifiers
CAS5123-13-7
IUPAC Name5,5-dimethyl-2-phenyl-1,3,2-dioxaborinane
Molecular FormulaC11H15BO2
InChI KeyFQMSNYZDFMWLGC-UHFFFAOYSA-N
SMILESCC1(C)COB(OC1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Solid
GC>=96.0 %
Infrared spectrumConforms
Appearance (Color)White
Melting point62°C to 67°C