Pyrene, 98%, Thermo Scientific Chemicals
Pyrene, 98%, Thermo Scientific Chemicals
Pyrene, 98%, Thermo Scientific Chemicals
Pyrene, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Pyrene, 98%, Thermo Scientific Chemicals
CAS: 129-00-0 | C16H10 | 202.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Packaging:
Glass bottle
Catalog number ACR180835000
Price (MYR)
2,413.00
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
2,413.00
Add to cart
Chemical Identifiers
CAS7114-03-6
IUPAC Namezinc(2+) bis[4-(dimethylamino)phenyl][4-(ethyldimethylazaniumyl)phenyl]methylium tetrachloride
Molecular FormulaC27H35Cl4N3Zn
InChI KeyZKOGXCSOLMZRAZ-UHFFFAOYSA-J
SMILES[Cl-].[Cl-].[Cl-].[Cl-].[Zn++].CC[N+](C)(C)C1=CC=C(C=C1)[C+](C1=CC=C(C=C1)N(C)C)C1=CC=C(C=C1)N(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
BSC CertificationPasses
Appearance (Color)Dark green to dark brown-red
Appearance (Form)Powder
Infrared spectrumConforms
Dye content>=80 %