Pyrene, 98%, Thermo Scientific Chemicals
Pyrene, 98%, Thermo Scientific Chemicals
Pyrene, 98%, Thermo Scientific Chemicals
Pyrene, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Pyrene, 98%, Thermo Scientific Chemicals
CAS: 129-00-0 | C16H10 | 202.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
500 g
Packaging:
Glass bottle
Catalog number ACR180831000
Price (MYR)
774.00
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
774.00
Add to cart
Chemical Identifiers
CAS91742-21-1
IUPAC Name1,1,1-trifluoro-N-trifluoromethanesulfonylmethanesulfonamide sodium
Molecular FormulaC2HF6NNaO4S2
InChI KeyQXZNUMVOKMLCEX-UHFFFAOYSA-N
SMILES[Na].FC(F)(F)S(=O)(=O)NS(=O)(=O)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Titration)≥94%
Appearance (Color)White