Phenol, 99+%, for biochemistry, loose crystals, Thermo Scientific Chemicals
Phenol, 99+%, for biochemistry, loose crystals, Thermo Scientific Chemicals
Phenol, 99+%, for biochemistry, loose crystals, Thermo Scientific Chemicals
Phenol, 99+%, for biochemistry, loose crystals, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Phenol, 99+%, for biochemistry, loose crystals, Thermo Scientific Chemicals
CAS: 108-95-2 | C6H6O | 94.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Glass bottle
Catalog number ACR180781000
Price (MYR)
639.00
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
639.00
Add to cart
Chemical Identifiers
CAS67-63-0
IUPAC Namepropan-2-ol
Molecular FormulaC3H8O
InChI KeyKFZMGEQAYNKOFK-UHFFFAOYSA-N
SMILESCC(C)O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
Water=<0.2 % (K.F.)
Color scale=<10 APHA
Carbonyl compounds=<0.002 % (Propionaldehyde)
GC>=99.5 %