Terephthalic acid, 99+%, Thermo Scientific Chemicals
Terephthalic acid, 99+%, Thermo Scientific Chemicals
Terephthalic acid, 99+%, Thermo Scientific Chemicals
Terephthalic acid, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Terephthalic acid, 99+%, Thermo Scientific Chemicals
CAS: 100-21-0 | C8H6O4 | 166.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
250 g
1 kg
Packaging:
Plastic bottle
Catalog number ACR180725000
Price (MYR)
447.00
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
447.00
Add to cart
Chemical Identifiers
CAS1344-28-1
IUPAC Namedialuminium(3+) trioxidandiide
Molecular FormulaAl2O3
InChI KeyPNEYBMLMFCGWSK-UHFFFAOYSA-N
SMILES[O--].[O--].[O--].[Al+3].[Al+3]
View more
SpecificationsSpecification SheetSpecification Sheet
pH8.5-10.5