Terephthalic acid, 99+%, Thermo Scientific Chemicals
Terephthalic acid, 99+%, Thermo Scientific Chemicals
Terephthalic acid, 99+%, Thermo Scientific Chemicals
Terephthalic acid, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Terephthalic acid, 99+%, Thermo Scientific Chemicals
CAS: 100-21-0 | C8H6O4 | 166.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
500 g
1 kg
Packaging:
Plastic bottle
Catalog number ACR180722500
Price (MYR)
375.00
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
375.00
Add to cart
Chemical Identifiers
CAS57-71-6
IUPAC Name(3E)-3-(hydroxyimino)butan-2-one
Molecular FormulaC4H7NO2
InChI KeyFSEUPUDHEBLWJY-HWKANZROSA-N
SMILESCC(=O)C(\C)=N\O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
GC>=97.5 %
Sulfated ash=<0.1 %
Solubility(5 % in water) Clear to slightly hazy
Appearance (Color)White to off-white