Terephthalic acid, 99+%, Thermo Scientific Chemicals
Terephthalic acid, 99+%, Thermo Scientific Chemicals
Terephthalic acid, 99+%, Thermo Scientific Chemicals
Terephthalic acid, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Terephthalic acid, 99+%, Thermo Scientific Chemicals
CAS: 100-21-0 | C8H6O4 | 166.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
250 g
500 g
Packaging:
Plastic bottle
Catalog number ACR180720010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS926-06-7
IUPAC Namepropan-2-yl methanesulfonate
Molecular FormulaC4H10O3S
InChI KeySWWHCQCMVCPLEQ-UHFFFAOYSA-N
SMILESCC(C)OS(C)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
GC>=98.5 %
Appearance (Color)Clear colorless
Appearance (Form)Liquid
Refractive index1.4180 to 1.4210 (20°C, 589 nm)