Dimethyl terephthalate, 99%, Thermo Scientific Chemicals
Dimethyl terephthalate, 99%, Thermo Scientific Chemicals
Dimethyl terephthalate, 99%, Thermo Scientific Chemicals
Dimethyl terephthalate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Dimethyl terephthalate, 99%, Thermo Scientific Chemicals
CAS: 120-61-6 | C10H10O4 | 194.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
Packaging:
Plastic bottle
Catalog number ACR180585000
Price (MYR)
554.00
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
554.00
Add to cart
Chemical Identifiers
CAS7439-89-6
IUPAC Nameiron
Molecular FormulaFe
InChI KeyXEEYBQQBJWHFJM-UHFFFAOYSA-N
SMILES[Fe]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Granules
Particle size0.5 to 1 mm
Total trace metal impurities=<100 ppm
Appearance (Color)Silver grey