tert-Butyl hydroperoxide, 70% Solution in water, Thermo Scientific Chemicals
tert-Butyl hydroperoxide, 70% Solution in water, Thermo Scientific Chemicals
tert-Butyl hydroperoxide, 70% Solution in water, Thermo Scientific Chemicals
tert-Butyl hydroperoxide, 70% Solution in water, Thermo Scientific Chemicals
Thermo Scientific Chemicals
tert-Butyl hydroperoxide, 70% Solution in water, Thermo Scientific Chemicals
CAS: 75-91-2 | C4H10O2 | 90.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
5 g
500 g
2.5 kg
Packaging:
Plastic bottle
Catalog number ACR180342500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS40877-09-6
IUPAC Name4-chloro-1-(4-chlorophenyl)butan-1-one
Molecular FormulaC10H10Cl2O
InChI KeyUKCHLVFIVJBCKE-UHFFFAOYSA-N
SMILESClCCCC(=O)C1=CC=C(Cl)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.5570-1.5610 @ 20°C
Appearance (Color)Clear colorless to yellow to orange
Assay (GC)≥96.0%