Petroleum ether, ACS reagent, boiling range 40-60°C, Thermo Scientific Chemicals
Petroleum ether, ACS reagent, boiling range 40-60°C, Thermo Scientific Chemicals
Petroleum ether, ACS reagent, boiling range 40-60°C, Thermo Scientific Chemicals
Petroleum ether, ACS reagent, boiling range 40-60°C, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Petroleum ether, ACS reagent, boiling range 40-60°C, Thermo Scientific Chemicals
CAS: 64742-49-0 | C6H14 | 86.178 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Packaging:
Glass Bottle
Catalog number ACR180210025
Price (MYR)
826.00
Add to cart
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
826.00
Add to cart
Chemical Identifiers
CAS149-91-7
IUPAC Name3,4,5-trihydroxybenzoic acid
Molecular FormulaC7H6O5
InChI KeyLNTHITQWFMADLM-UHFFFAOYSA-N
SMILESOC(=O)C1=CC(O)=C(O)C(O)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream or pale yellow
FormPowder
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
Assay (Silylated GC)≥97.5 %