p-Dithiane-2,5-diol, 96%, Thermo Scientific Chemicals
p-Dithiane-2,5-diol, 96%, Thermo Scientific Chemicals
p-Dithiane-2,5-diol, 96%, Thermo Scientific Chemicals
p-Dithiane-2,5-diol, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
p-Dithiane-2,5-diol, 96%, Thermo Scientific Chemicals
CAS: 40018-26-6 | C4H8O2S2 | 152.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
Packaging:
Plastic bottle
Catalog number ACR179932500
Price (MYR)
1,479.00
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
1,479.00
Add to cart
Chemical Identifiers
CAS51512-09-5
IUPAC Name2-(2-chlorophenyl)acetyl chloride
Molecular FormulaC8H6Cl2O
InChI KeyWIHSAOYVGKVRJX-UHFFFAOYSA-N
SMILESClC(=O)CC1=CC=CC=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Refractive index1.5480 to 1.5500 (20°C, 589 nm)
Titration RCOCl>=96.0 %
Appearance (Form)Liquid
Appearance (Color)Clear yellow