2'-Fluoroacetophenone, 97%
2'-Fluoroacetophenone, 97%
2'-Fluoroacetophenone, 97%
2'-Fluoroacetophenone, 97%
Thermo Scientific Chemicals
2'-Fluoroacetophenone, 97%
CAS: 445-27-2 | C8H7FO | 138.14 g/mol
Have Questions?
Catalog number ACR179780250
Price (MYR)
1,230.00
Add to cart
Request bulk or custom format
Price (MYR)
1,230.00
Add to cart
Chemical Identifiers
CAS136-17-4
IUPAC NameN1-phenylbenzene-1,2,4-triamine
Molecular FormulaC12H13N3
InChI KeyVOSLIUIVGWBSOK-UHFFFAOYSA-N
SMILESNC1=CC=C(NC2=CC=CC=C2)C(N)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
Identification (FTIR)Conforms
Melting Point (clear melt)126.0-133.0?C
FormPowder
Appearance (Color)Brown or purple to black