2'-Chloroacetophenone, 97%, Thermo Scientific Chemicals
2'-Chloroacetophenone, 97%, Thermo Scientific Chemicals
2'-Chloroacetophenone, 97%, Thermo Scientific Chemicals
2'-Chloroacetophenone, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2'-Chloroacetophenone, 97%, Thermo Scientific Chemicals
CAS: 2142-68-9 | C8H7ClO | 154.6 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number ACR179770250
Price (MYR)
619.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
619.00
Add to cart
Chemical Identifiers
CAS7758-19-2
IUPAC Namesodium chlorite
Molecular FormulaClNaO2
InChI KeyUKLNMMHNWFDKNT-UHFFFAOYSA-M
SMILES[Na+].[O-][Cl]=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Titration Iodometric>=75 %
Appearance (Form)Powder