Naphazoline hydrochloride, 99%, Thermo Scientific Chemicals
Naphazoline hydrochloride, 99%, Thermo Scientific Chemicals
Naphazoline hydrochloride, 99%, Thermo Scientific Chemicals
Naphazoline hydrochloride, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Naphazoline hydrochloride, 99%, Thermo Scientific Chemicals
CAS: 550-99-2 | C14H14N2·ClH | 246.74 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Plastic bottle
Catalog number ACR179661000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS78-10-4
IUPAC Nametetraethyl silicate
Molecular FormulaC8H20O4Si
InChI KeyBOTDANWDWHJENH-UHFFFAOYSA-N
SMILESCCO[Si](OCC)(OCC)OCC
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
GC>=97.5 %
Color scale=<20 APHA
Refractive index1.3820 to 1.3840 (20°C, 589 nm)
GC=<1 % Ethanol