2-(Aminomethyl)-1-ethylpyrrolidine, 95%, Thermo Scientific Chemicals
2-(Aminomethyl)-1-ethylpyrrolidine, 95%, Thermo Scientific Chemicals
2-(Aminomethyl)-1-ethylpyrrolidine, 95%, Thermo Scientific Chemicals
2-(Aminomethyl)-1-ethylpyrrolidine, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-(Aminomethyl)-1-ethylpyrrolidine, 95%, Thermo Scientific Chemicals
CAS: 26116-12-1 | C7H16N2 | 128.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
Packaging:
Glass bottle
Catalog number ACR179480100
Price (MYR)
1,089.00
Add to cart
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,089.00
Add to cart
Chemical Identifiers
CAS66-99-9
IUPAC Namenaphthalene-2-carbaldehyde
Molecular FormulaC11H8O
InChI KeyPJKVFARRVXDXAD-UHFFFAOYSA-N
SMILESO=CC1=CC=C2C=CC=CC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Off-white to yellow to light beige
Infrared spectrumConforms
GC>=97.5 %
Appearance (Form)Powder and/or chunks
Melting point58°C to 62°C